Vidarabine 99%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Adenine 9-β-D-arabinofuranoside, 9-β-D-Arabinofuranosyladenine, (2R,3S,4S,5R)-2-(6-Amino-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol, 9-β-D-Arabinofuranosyladenine, Ara-A, Vidarabine
J65076.06
J65076.14
J65076.06EA
60.2
EUR
J65076.06
J65076.14
Vidarabine 99%
Vidarabine
Purity: 99%
CAS No.: 5536-17-4
Molecular Formula: C10H13N5O4
Formula Weight: 267.24
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White to off-white
Beilstein Reference: 624881
MDL No.: MFCD00065471
Formula:
C₁₀H₁₃N₅O₄ MW: 267,24 g/mol |
MDL Number:
MFCD00065471 CAS Number: 5536-17-4 EINECS: 226-893-9 Merck Index: 14,9976 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...