Flumequine 98%

Supplier: Thermo Scientific

Synonyms: 9-Fluoro-5-methyl-1-oxo-6,7-dihydro-1H,5H-pyrido[3,2,1-ij]quinoline-2-carboxylic acid, 9-Fluoro-1,5,6,7-tetrahydro-5-methyl-1-oxopyrido[3,2,1-ij]quinoline-2-carboxylic acid

J66332.03 J66332.06
J66332.03EA 64.2 EUR
J66332.03 J66332.06
Flumequine 98%
Flumequine

Purity: 98% CAS No.: 42835-25-6 Molecular Formula: C14H12FNO3 Formula Weight: 261.25 Flash Point: 110°(230°F) Physical Form: Crystalline powder Appearance: White Melting Point: 253-255° Solubility: Soluble in DMSO and dilute alkali hydroxides. Insoluble in water Beilstein Reference: 490724 MDL No.: MFCD00079298


Applications: Inhibits topoisomerases in gram negative bacteria.

Formula: C₁₄H₁₂FNO₃
MW: 261,25 g/mol
Melting Pt: 253…255 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00079298
CAS Number: 42835-25-6
EINECS: 255-962-6
Merck Index: 14,4137

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR