Phencyclone 97%
Supplier: Thermo Fisher Scientific
L00108.03
L00108.06
L00108.03EA
63.2
EUR
L00108.03
L00108.06
Phencyclone 97%
Phencyclone
Purity: 97%
CAS No.: 5660-91-3
Molecular Formula: C29H18O
Formula Weight: 382.46
Melting Point: 245-249°
Beilstein Reference: 2061817
MDL No.: MFCD00051451
Formula:
C₂₉H₁₈O Melting Pt: 245…249 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051451 CAS Number: 5660-91-3 EINECS: 227-112-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...