(2-Benzimidazolyl)acetonitrile 99%
Supplier: Thermo Scientific
|
Warning
|
L00827.04
L00827.09
L00827.18
L00827.04EA
21.6
EUR
L00827.04
L00827.09
L00827.18
(2-Benzimidazolyl)acetonitrile 99%
(2-Benzimidazolyl)acetonitrile
Purity: 99%
CAS No.: 4414-88-4
Molecular Formula: C9H7N3
Formula Weight: 157.18
Odour: Odourless
Melting Point: 206-211°
Beilstein Reference: 128461
MDL No.: MFCD00005601
Formula:
C₉H₇N₃ MW: 157,18 g/mol Melting Pt: 206…211 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005601 CAS Number: 4414-88-4 EINECS: 224-574-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...