Ethyl-3-dimethylaminopropionate 97%
Supplier: Thermo Fisher Scientific
|
Warning
|
Purity: 97%
CAS No.: 20120-21-2
Molecular Formula: (CH3)2NCH2CH2CO2CH2CH3
Formula Weight: 145.20
Flash Point: 58°(136°F)
Refractive Index: 1.4190
Odour: Amine-like
Boiling Point: 56-57°/12mm
Beilstein Reference: 1702477
UN No.: UN1993
MDL No.: MFCD00015166
Formula:
(CH₃)₂NCH₂CH₂CO₂CH₂CH₃ Boiling Pt: 56…57 °C (12 mmHg) Density: 0,915 Flash Pt: 58 °C (136 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00015166 CAS Number: 20120-21-2 EINECS: 243-524-7 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...