Pyrrolidin-1-ylacetonitrile 97%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
2-(Pyrrolidin-1-yl)acetonitrile
L01737.03
L01737.06
L01737.03EA
0
EUR
L01737.03
L01737.06
Pyrrolidin-1-ylacetonitrile 97%
Pyrrolidin-1-ylacetonitrile
Purity: 97%
CAS No.: 29134-29-0
Molecular Formula: C6H10N2
Formula Weight: 110.16
Refractive Index: 1.4563
Beilstein Reference: 106157
UN No.: UN3276
MDL No.: MFCD00014098
Formula:
C₆H₁₀N₂ MW: 110,16 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00014098 CAS Number: 29134-29-0 EINECS: 249-463-2 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...