Norcantharidin ≥98%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
exo-7-Oxabicyclo[2.2.1]heptane-2,3-dicarboxylic Anhydride, rel-(3aR,4R,7S,7aS)-Hexahydro-4,7-epoxyisobenzofuran-1,3-dione, exo-3,6-Epoxyhexahydrophthalic Anhydride
L02947.06
L02947.14
L02947.06EA
0
EUR
L02947.06
L02947.14
Norcantharidin ≥98%
Norcantharidin
Purity: 98+%
CAS No.: 29745-04-8
Molecular Formula: C8H8O4
Formula Weight: 168.15
Melting Point: 111-116°
Beilstein Reference: 84287
MDL No.: MFCD00213361
Formula:
C₈H₈O₄ Melting Pt: 111…116 °C Storage Temperature: Ambient |
MDL Number:
MFCD00213361 CAS Number: 29745-04-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...