Monoethyl glutarate 97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
5-Ethoxy-5-oxopentanoic acid, Glutaric Acid Monoethyl Ester
L03941.06
L03941.14
L03941.06EA
85.4
EUR
L03941.06
L03941.14
Monoethyl glutarate 97%
Monoethyl glutarate
Purity: 97%
CAS No.: 1070-62-8
Molecular Formula: C7H12O4
Formula Weight: 160.17
Refractive Index: 1.4365
Boiling Point: 118-120°/1.5mm
Beilstein Reference: 1766397
MDL No.: MFCD00014379
Formula:
C₇H₁₂O₄ MW: 160,17 g/mol Boiling Pt: 118…120 °C (1,5 mmHg) Storage Temperature: Ambient |
MDL Number:
MFCD00014379 CAS Number: 1070-62-8 EINECS: 213-977-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...