Ethyl hydrogen sebacate 98%
Supplier: Thermo Scientific
|
Warning
|
L04134.06
L04134.14
L04134.06EA
120
EUR
L04134.06
L04134.14
Ethyl hydrogen sebacate 98%
Ethyl hydrogen sebacate
Purity: 98%
CAS No.: 693-55-0
Molecular Formula: CH3CH2O2C(CH2)8CO2H
Formula Weight: 230.30
Melting Point: 35-38°
Beilstein Reference: 1782569
MDL No.: MFCD00014387
Formula:
CH₃CH₂O₂C(CH₂)₈CO₂H MW: 230,30 g/mol Melting Pt: 35…38 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014387 CAS Number: 693-55-0 EINECS: 211-753-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...