3-(4-Methylpiperazin-1-yl)propylamine 98%

Supplier: Thermo Fisher Scientific
Danger

L04876.03 L04876.06 L04876.14
L04876.03EA 70.9 EUR
L04876.03 L04876.06 L04876.14
3-(4-Methylpiperazin-1-yl)propylamine 98%
3-(4-Methylpiperazin-1-yl)propylamine

Purity: 98% CAS No.: 4572-03-6 Molecular Formula: C8H19N3 Formula Weight: 157.26 Refractive Index: 1.4820 Boiling Point: 113-114°/13mm Beilstein Reference: 105964 UN No.: UN2735 MDL No.: MFCD00014616

Formula: C₈H₁₉N₃
Boiling Pt: 113…114 °C (13 mmHg)
Density: 0,924
Storage Temperature: Ambient
MDL Number: MFCD00014616
CAS Number: 4572-03-6
EINECS: 224-954-4
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR