6-Methoxy-2(3H)-benzoxazolone ≥98%
Supplier: Thermo Scientific
Synonyms:
Coixol, 6-Methoxybenzo[d]oxazol-2(3H)-one
L05349.03
L05349.MD
L05349.03EA
202
EUR
L05349.03
L05349.MD
6-Methoxy-2(3H)-benzoxazolone ≥98%
6-Methoxy-2(3H)-benzoxazolone
Purity: 98+%
CAS No.: 532-91-2
Molecular Formula: C8H7NO3
Formula Weight: 165.15
Melting Point: 152-156°
Beilstein Reference: 141041
MDL No.: MFCD00051726
Formula:
C₈H₇NO₃ MW: 165,15 g/mol Melting Pt: 152…156 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051726 CAS Number: 532-91-2 |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...