α-Cyclohexylphenylacetonitrile 98%
Supplier: Thermo Fisher Scientific
|
Danger
|
L05350.09
L05350.18
L05350.09EA
24.6
EUR
L05350.09
L05350.18
α-Cyclohexylphenylacetonitrile 98%
alpha-Cyclohexylphenylacetonitrile
Purity: 98%
CAS No.: 3893-23-0
Molecular Formula: C14H17N
Formula Weight: 199.30
Melting Point: 52-56°
Boiling Point: 137-140°/3mm
Beilstein Reference: 2051115
UN No.: UN3439
MDL No.: MFCD00019362
Formula:
C₁₄H₁₇N Boiling Pt: 137…140 °C (3 mmHg) Melting Pt: 52…56 °C Storage Temperature: Ambient |
MDL Number:
MFCD00019362 CAS Number: 3893-23-0 EINECS: 223-442-8 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...