1-(p-Tolyl)piperazine dihydrochloride 97%

Supplier: Thermo Fisher Scientific
Warning

L05432.06 L05432.14
L05432.06EA 0 EUR
L05432.06 L05432.14
1-(p-Tolyl)piperazine dihydrochloride 97%
1-(p-Tolyl)piperazine dihydrochloride

Purity: 97% CAS No.: 13078-14-3 Molecular Formula: C11H16N2.2HCl Formula Weight: 249.19 Melting Point: 214-216° Beilstein Reference: 5639546 MDL No.: MFCD00035286

Formula: C₁₁H₁₆N₂·2HCl
MW: 249.19 g/mol
Melting Pt: 214…216 °C
Storage Temperature: Ambient
MDL Number: MFCD00035286
CAS Number: 13078-14-3
EINECS: 235-975-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR