5-Cyclopropyl-1,3,4-thiadiazol-2-amine 98%
Supplier: Thermo Scientific
L05778.03
L05778.06
L05778.03EA
48.3
EUR
L05778.03
L05778.06
5-Cyclopropyl-1,3,4-thiadiazol-2-amine 98%
5-Cyclopropyl-1,3,4-thiadiazol-2-amine
Purity: 98%
CAS No.: 57235-50-4
Molecular Formula: C5H7N3S
Formula Weight: 141.20
Melting Point: 212-216°
Beilstein Reference: 124356
MDL No.: MFCD00051649
Formula:
C₅H₇N₃S Melting Pt: 212…216 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051649 CAS Number: 57235-50-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...