3-Indazolinone 97%

Supplier: Thermo Scientific

Synonyms: 1,2-Dihydro-3H-indazol-3-one

L05860.06 L05860.14 L05860.22
L05860.06EA 77.5 EUR
L05860.06 L05860.14 L05860.22
3-Indazolinone 97%
3-Indazolinone

Purity: 97% CAS No.: 7364-25-2 Molecular Formula: C7H6N2O Formula Weight: 134.14 Melting Point: 248-252° Beilstein Reference: 3733 MDL No.: MFCD00005685

Formula: C₇H₆N₂O
MW: 134,14 g/mol
Melting Pt: 248…252 °C
Storage Temperature: Ambient
MDL Number: MFCD00005685
CAS Number: 7364-25-2
EINECS: 230-904-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR