5-Amino-3-(2-thienyl)-1H-pyrazole 98%

Supplier: Thermo Scientific
Warning

Synonyms: 5-Thien-2-yl-1H-pyrazol-3-amine

L06752.03 L06752.06
L06752.03EA 43 EUR
L06752.03 L06752.06
5-Amino-3-(2-thienyl)-1H-pyrazole 98%
5-Amino-3-(2-thienyl)-1H-pyrazole

Purity: 98% CAS No.: 96799-03-0 Molecular Formula: C7H7N3S Formula Weight: 165.21 Melting Point: 101-103° Beilstein Reference: 4399374 MDL No.: MFCD00051815

Formula: C₇H₇N₃S
MW: 165,21 g/mol
Melting Pt: 101…103 °C
Storage Temperature: Ambient
MDL Number: MFCD00051815
CAS Number: 96799-03-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR