3-Acetoxy-2-butanone 98%

Supplier: Thermo Fisher Scientific
Warning

L06927.06 L06927.14
L06927.06EA 37.4 EUR
L06927.06 L06927.14
3-Acetoxy-2-butanone 98%
3-Acetoxy-2-butanone

Purity: 98% CAS No.: 4906-24-5 Molecular Formula: C6H10O3 Formula Weight: 130.14 Flash Point: 66°(150°F) Refractive Index: 1.4135 Boiling Point: 154-155° Beilstein Reference: 1756882 MDL No.: MFCD00051782

Formula: C₆H₁₀O₃
Boiling Pt: 154…155 °C
Density: 1,025
Flash Pt: 66 °C (150 °F)
Storage Temperature: Ambient
MDL Number: MFCD00051782
CAS Number: 4906-24-5

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR