3-Bromobenzanthrone ≥85%, tech.
Supplier: Thermo Fisher Scientific
|
Danger
|
L07422.14
L07422.22
L07422.14EA
61.6
EUR
L07422.14
L07422.22
3-Bromobenzanthrone ≥85%, tech.
3-Bromobenzanthrone
Purity: 85%
CAS No.: 81-96-9
Molecular Formula: C17H9BrO
Formula Weight: 309.17
Melting Point: 164-168°
Beilstein Reference: 1975512
MDL No.: MFCD00021094
Formula:
C₁₇H₉BrO Melting Pt: 164…168 °C Storage Temperature: Ambient |
MDL Number:
MFCD00021094 CAS Number: 81-96-9 EINECS: 201-390-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...