Ethyl-1-benzyl-3-oxopiperidine-4-carboxylate hydrochloride 90%, tech.
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Ethyl 1-Benzyl-3-oxo-4-piperidinecarboxylate HCl, Ethyl N-benzyl-3-oxo-4-piperidinecarboxylate hydrochloride, Ethyl 1-benzyl-3-oxopiperidine-4-carboxylate hydrochloride
L07472.03
L07472.06
L07472.03EA
31.5
EUR
L07472.03
L07472.06
Ethyl-1-benzyl-3-oxopiperidine-4-carboxylate hydrochloride 90%, tech.
Ethyl-1-benzyl-3-oxopiperidine-4-carboxylate hydrochloride
Purity: 90%
CAS No.: 52763-21-0
Molecular Formula: C15H19NO3.HCl
Formula Weight: 297.78
Melting Point: ca 158° dec.
Beilstein Reference: 3749159
MDL No.: MFCD00012792
Formula:
C₁₅H₂₀ClNO₃ MW: 297,78 g/mol Melting Pt: ∼ 158 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00012792 CAS Number: 52763-21-0 EINECS: 258-162-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...