tert-Butylsulphonylacetonitrile 98%
Supplier: Thermo Fisher Scientific
|
Danger
|
L07525.03
L07525.06
L07525.03EA
75.8
EUR
L07525.03
L07525.06
tert-Butylsulphonylacetonitrile 98%
tert-Butylsulphonylacetonitrile
Purity: 98%
CAS No.: 81536-18-7
Molecular Formula: C6H11NO2S
Formula Weight: 161.22
Melting Point: 142-144°
Beilstein Reference: 4370746
UN No.: UN3439
MDL No.: MFCD00051622
Formula:
C₆H₁₁NO₂S Melting Pt: 142…144 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051622 CAS Number: 81536-18-7 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...