Ethyl-3-phenoxypropionate 98%
Supplier: Thermo Fisher Scientific
Purity: 98%
CAS No.: 22409-91-2
Molecular Formula: C6H5OCH2CH2CO2CH2CH3
Formula Weight: 194.23
Flash Point:
Formula:
C₆H₅OCH₂CH₂CO₂CH₂CH₃ Boiling Pt: 141…143 °C (11 mmHg) Melting Pt: 22…24 °C Density: 1,084 Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00026919 CAS Number: 22409-91-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...