Ethyl acetamidocyanoacetate 98%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
Ethyl 2-acetamido-2-cyanoacetate
L07674.09
L07674.04
L07674.09EA
77.5
EUR
L07674.09
L07674.04
Ethyl acetamidocyanoacetate 98%
Ethyl acetamidocyanoacetate
Purity: 98%
CAS No.: 4977-62-2
Molecular Formula: C7H10N2O3
Formula Weight: 170.17
Melting Point: 128-130°
Beilstein Reference: 879845
MDL No.: MFCD00009133
Formula:
C₇H₁₀N₂O₃ MW: 170,168 g/mol Melting Pt: 128…130 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009133 CAS Number: 4977-62-2 EINECS: 225-624-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...