N-Carbobenzoxy-L-leucine 98% (dry weight) may cont. up to ca 5% solvent
Supplier: Thermo Fisher Scientific
L08471.06
L08471.14
L08471.06EA
0
EUR
L08471.06
L08471.14
N-Carbobenzoxy-L-leucine 98% (dry weight) may cont. up to ca 5% solvent
N-Carbobenzoxy-L-leucine
Purity: 98+%
CAS No.: 2018-66-8
Molecular Formula: C14H19NO4
Formula Weight: 265.30
Storage Temperature: 2°C to 8°C
Flash Point:
Formula:
C₁₄H₁₉NO₄ Density: 1,00 Flash Pt: >110 °C (230 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00026494 CAS Number: 2018-66-8 EINECS: 217-960-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...