5-Isocyanato-1,2,3-trimethoxybenzene 97%
Supplier: Thermo Fisher Scientific
|
Danger
|
Synonyms:
3,4,5-Trimethoxyphenyl isocyanate
L09775.03
L09775.06
L09775.03EA
22.8
EUR
L09775.03
L09775.06
5-Isocyanato-1,2,3-trimethoxybenzene 97%
5-Isocyanato-1,2,3-trimethoxybenzene
Purity: 97%
CAS No.: 1016-19-9
Molecular Formula: C10H11NO4
Formula Weight: 209.20
Storage Temperature: 2°C to 8°C
Flash Point:
Formula:
C₁₀H₁₁NO₄ Boiling Pt: 102…110 °C (0,4 mmHg) Melting Pt: 39…41 °C Flash Pt: >110 °C (230 °F) |
MDL Number:
MFCD00013861 CAS Number: 1016-19-9 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...