3-Methylquinoline-N-oxide 97%
Supplier: Thermo Fisher Scientific
L10635.03
L10635.MD
L10635.03EA
59.5
EUR
L10635.03
L10635.MD
3-Methylquinoline-N-oxide 97%
3-Methylquinoline-N-oxide
Purity: 97%
CAS No.: 1873-55-8
Molecular Formula: C10H9NO
Formula Weight: 159.19
Melting Point: 88-89°
Beilstein Reference: 118125
MDL No.: MFCD00082650
Formula:
C₁₀H₉NO Melting Pt: 88…89 °C Storage Temperature: Ambient |
MDL Number:
MFCD00082650 CAS Number: 1873-55-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...