1-Pyrenecarboxaldehyde 99%
Supplier: Thermo Fisher Scientific
|
Warning
|
L10835.06
L10835.14
L10835.06EA
82.3
EUR
L10835.06
L10835.14
1-Pyrenecarboxaldehyde 99%
1-Pyrenecarboxaldehyde
Purity: 99%
CAS No.: 3029-19-4
Molecular Formula: C17H10O
Formula Weight: 230.27
Melting Point: 125-128°
Beilstein Reference: 1874889
MDL No.: MFCD00004139
Formula:
C₁₇H₁₀O Melting Pt: 125…128 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004139 CAS Number: 3029-19-4 EINECS: 221-196-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...