Thymolphthalein complexon

Supplier: Thermo Fisher Scientific

L10962.03 L10962.06
L10962.03EA 36 EUR
L10962.03 L10962.06
Thymolphthalein complexon
Thymolphthalein complexon

CAS No.: 1913-93-5 Molecular Formula: C38H44N2O12 Formula Weight: 720.78 Beilstein Reference: 1339087 MDL No.: MFCD00001619

Formula: C₃₈H₄₄N₂O₁₂
MW: 720.77 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00001619
CAS Number: 1913-93-5
EINECS: 217-627-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR