Propargyl methacrylate 98% stabilised
Supplier: Thermo Fisher Scientific
|
Warning
|
Stabilisation:
Stabilised with 4-methoxyphenol (MEHQ) 250 ppm
L11556.09
L11556.18
L11556.09EA
119
EUR
L11556.09
L11556.18
Propargyl methacrylate 98% stabilised
Propargyl methacrylate
Stab. with 250ppm 4-methoxyphenol
Purity: 98%
CAS No.: 13861-22-8
Molecular Formula: C7H8O2
Formula Weight: 124.14
Storage Temperature: 2°C to 8°C
Flash Point: 48°(118°F)
Refractive Index: 1.4500
Boiling Point: 149-151°
Beilstein Reference: 906850
UN No.: UN3272
MDL No.: MFCD00048123
Formula:
C₇H₈O₂ Boiling Pt: 149…151 °C Density: 0,986 Flash Pt: 48 °C (118 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00048123 CAS Number: 13861-22-8 EINECS: 237-599-5 UN: 3272 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...