4-Vinylphenyl acetate 95% stabilised
Supplier: Thermo Fisher Scientific
|
Danger
|
Synonyms:
Acetic Acid 4-Vinylphenyl Ester, 4-Acetoxystyrene
L11601.09
L11601.04
L11601.09EA
71.2
EUR
L11601.09
L11601.04
4-Vinylphenyl acetate 95% stabilised
4-Vinylphenyl acetate
Purity: 95%
CAS No.: 2628-16-2
Molecular Formula: C10H10O2
Formula Weight: 162.19
Flash Point: 87°(188°F)
Refractive Index: 1.5380
Melting Point: 7-8°
Boiling Point: 259-260°
Beilstein Reference: 1862793
MDL No.: MFCD00075734
Formula:
C₁₀H₁₀O₂ Boiling Pt: 259…260 °C Melting Pt: 7…8 °C Density: 1,060 Flash Pt: 87 °C (188 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00075734 CAS Number: 2628-16-2 EINECS: 434-600-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...