(+/-)-1-(4-Bromophenyl)ethyl isocyanate 97%

Supplier: Thermo Scientific
Danger

L11836.03 L11836.06
L11836.03EA 0 EUR
L11836.03 L11836.06
(+/-)-1-(4-Bromophenyl)ethyl isocyanate 97%
(+/-)-1-(4-Bromophenyl)ethyl isocyanate

Purity: 97% CAS No.: 207974-15-0 Molecular Formula: C9H8BrNO Formula Weight: 226.08 Storage Temperature: 2°C to 8°C Refractive Index: 1.5565 Boiling Point: 145-147°/7mm UN No.: UN2206 MDL No.: MFCD00041411

Formula: C₉H₈BrNO
MW: 226,08 g/mol
Boiling Pt: 145…147 °C (7 mmHg)
MDL Number: MFCD00041411
CAS Number: 207974-15-0
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR