N-Acryloylsarcosine methyl ester 97% stabilised

Supplier: Thermo Fisher Scientific
Warning


Stabilisation: Stabilised with 4-methoxyphenol (MEHQ)

L13128.09 L13128.04
L13128.09EA 0 EUR
L13128.09 L13128.04
N-Acryloylsarcosine methyl ester 97% stabilised
N-Acryloylsarcosine methyl ester

Stab. with 250ppm 4-methoxyphenol


Purity: 97% CAS No.: 72065-23-7 Molecular Formula: C7H11NO3 Formula Weight: 157.17 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Refractive Index: 1.4770 Boiling Point: 96°/0.4mm Beilstein Reference: 4964477 MDL No.: MFCD00151792

Formula: C₇H₁₁NO₃
Boiling Pt: 96 °C (0,4 mmHg)
Density: 1,02
Flash Pt: >110 °C (230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00151792
CAS Number: 72065-23-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR