Cyclopropanemethylamine hydrochloride 98%

Supplier: Thermo Scientific
Warning

Synonyms: (Aminomethyl)cyclopropane hydrochloride

L13353.03 L13353.06
L13353.03EA 50.1 EUR
L13353.03 L13353.06
Cyclopropanemethylamine hydrochloride 98%
Cyclopropanemethylamine hydrochloride

Purity: 98% CAS No.: 7252-53-1 Molecular Formula: C4H9N.HCl Formula Weight: 107.58 Melting Point: 202-205° Beilstein Reference: 3906936 MDL No.: MFCD00012544

Formula: C₄H₉N·HCl
MW: 107,58 g/mol
Melting Pt: 202…205 °C
Storage Temperature: Ambient
MDL Number: MFCD00012544
CAS Number: 7252-53-1
EINECS: 230-668-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR