5-Oxo-5-(2-thienyl)valeric acid 97%
Supplier: Thermo Scientific
|
Warning
|
L13362.03
L13362.06
L13362.03EA
33
EUR
L13362.03
L13362.06
5-Oxo-5-(2-thienyl)valeric acid 97%
5-Oxo-5-(2-thienyl)valeric acid
Purity: 97%
CAS No.: 22971-62-6
Molecular Formula: C9H10O3S
Formula Weight: 198.24
Melting Point: 94-97°
Beilstein Reference: 141775
MDL No.: MFCD00151848
Formula:
C₉H₁₀O₃S MW: 198,24 g/mol Melting Pt: 94…97 °C Storage Temperature: Ambient |
MDL Number:
MFCD00151848 CAS Number: 22971-62-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...