3',4',7,8-Tetramethoxyflavone 97%
Supplier: Thermo Scientific
L13829.MF
L13829.MD
L13829.MFEA
45.4
EUR
L13829.MF
L13829.MD
3',4',7,8-Tetramethoxyflavone 97%
3',4',7,8-Tetramethoxyflavone
Purity: 97%
CAS No.: 65548-55-2
Molecular Formula: C19H18O6
Formula Weight: 342.35
Melting Point: 206-208°
Beilstein Reference: 330821
MDL No.: MFCD00143001
Formula:
C₁₉H₁₈O₆ MW: 342,35 g/mol Melting Pt: 206…208 °C Storage Temperature: Ambient |
MDL Number:
MFCD00143001 CAS Number: 65548-55-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...