Cycloheptanemethylamine ≥98%
Supplier: Thermo Fisher Scientific
|
Danger
|
L14659.03
L14659.06
L14659.03EA
62.6
EUR
L14659.03
L14659.06
Cycloheptanemethylamine ≥98%
Cycloheptanemethylamine
Purity: 98+%
CAS No.: 4448-77-5
Molecular Formula: C8H17N
Formula Weight: 127.23
Flash Point: 70°(158°F)
Refractive Index: 1.4740
Boiling Point: 98-101°/35mm
Beilstein Reference: 2958734
UN No.: UN2735
MDL No.: MFCD00021674
Formula:
C₈H₁₇N Boiling Pt: 98…101 °C (35 mmHg) Density: 0,884 Flash Pt: 70 °C (158 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00021674 CAS Number: 4448-77-5 EINECS: 224-692-0 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...