2,4-Dihydroxy-3-nitropyridine 99%

Supplier: Thermo Scientific
Warning

Synonyms: 3-Nitropyridine-2,4-diol

L14933.03
L14933.03EA 24.6 EUR
L14933.03
2,4-Dihydroxy-3-nitropyridine 99%
2,4-Dihydroxy-3-nitropyridine

Purity: 99% CAS No.: 89282-12-2 Molecular Formula: C5H4N2O4 Formula Weight: 156.10 Melting Point: ca 265° dec. Beilstein Reference: 149558 MDL No.: MFCD01075671

Formula: C₅H₄N₂O₄
MW: 156,10 g/mol
Melting Pt: ∼ 265 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD01075671
CAS Number: 89282-12-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR