Sodium vinylsulphonate ∼25% in aqueous solution, Technical Grade

Supplier: Thermo Fisher Scientific
Warning

L15356.AE L15356.AP
L15356.AEEA 38.4 EUR
L15356.AE L15356.AP
Sodium vinylsulphonate ∼25% in aqueous solution, Technical Grade
Sodium vinylsulphonate

CAS No.: 3039-83-6 Molecular Formula: CH2=CHSO3Na Formula Weight: 130.10 Refractive Index: 1.3770 MDL No.: MFCD00007520

Formula: C₂H₄O₃S
Density: 1,180
Storage Temperature: Ambient
MDL Number: MFCD00007520
CAS Number: 3039-83-6
EINECS: 221-242-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR