3-Furaldehyde diethyl acetal 98%
Supplier: Thermo Scientific
|
Danger
|
L15519.06
L15519.06EA
133
EUR
L15519.06
3-Furaldehyde diethyl acetal 98%
3-Furaldehyde diethyl acetal
Purity: 98%
CAS No.: 216144-29-5
Molecular Formula: C9H14O3
Formula Weight: 170.21
Refractive Index: 1.4440
MDL No.: MFCD01318151
Formula:
C₉H₁₄O₃ MW: 170,21 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01318151 CAS Number: 216144-29-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...