4,5-Dimethoxy-o-phenylenediamine dihydrochloride 98%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
4,5-Dimethoxybenzene-1,2-diamine dihydrochloride
L15626.MD
L15626.03
L15626.MDEA
123
EUR
L15626.MD
L15626.03
4,5-Dimethoxy-o-phenylenediamine dihydrochloride 98%
4,5-Dimethoxy-o-phenylenediamine dihydrochloride
Purity: 98%
CAS No.: 131076-14-7
Molecular Formula: C8H12N2O2.2HCl
Formula Weight: 241.12
Melting Point: ca 265° dec.
Beilstein Reference: 3714004
UN No.: UN2811
MDL No.: MFCD00190674
Formula:
C₈H₁₂N₂O₂·2HCl MW: 241,12 g/mol Melting Pt: ∼ 265 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00190674 CAS Number: 131076-14-7 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...