2-(4-Aminophenyl)ethylamine 98%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 4-(2-Aminoethyl)aniline, 1-Amino-2-(4-aminophenyl)ethane, 4-Aminophenethylamine

L15782.03 L15782.06
L15782.03EA 0 EUR
L15782.03 L15782.06
2-(4-Aminophenyl)ethylamine 98%
2-(4-Aminophenyl)ethylamine

Purity: 98% CAS No.: 13472-00-9 Molecular Formula: C8H12N2 Formula Weight: 136.20 Flash Point: 110°(230°F) Refractive Index: 1.5910 Melting Point: 28-31° Boiling Point: 103°/0.3mm Beilstein Reference: 1099913 UN No.: UN3259 MDL No.: MFCD00008194

Formula: C₈H₁₂N₂
Boiling Pt: 103 °C (0,3 mmHg)
Melting Pt: 28…31 °C
Density: 1,057
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008194
CAS Number: 13472-00-9
EINECS: 236-739-2
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR