Amberlite® XAD-1180 ion exchange resin

Supplier: Thermo Scientific

L16097.22 L16097.36
L16097.22EA 41.4 EUR
L16097.22 L16097.36
Amberlite® XAD-1180 ion exchange resin
Amberlite® XAD-1180

CAS No.: 97396-56-0 MDL No.: MFCD00145830

Storage Temperature: Ambient
MDL Number: MFCD00145830
CAS Number: 97396-56-0
EINECS: 000-000-0
Merck Index: 14,382

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR