1H,1H,11H-Eicosafluoroundecyl acrylate ≥85% stabilised, tech.
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
1H,1H,11H-Perfluoroundecyl acrylate
Stabilisation:
Stabilised with 4-methoxyphenol (MEHQ) 50 ppm
L16488.06
L16488.14
L16488.06EA
49.3
EUR
L16488.06
L16488.14
1H,1H,11H-Eicosafluoroundecyl acrylate ≥85% stabilised, tech.
1H,1H,11H-Eicosafluoroundecyl acrylate
Stab. with ca 50ppm 4-methoxyphenol
Purity: 85%
CAS No.: 4998-38-3
Molecular Formula: H2C=CHCO2CH2(CF2)10H
Formula Weight: 586.17
Storage Temperature: 2°C to 8°C
Melting Point: 44-53°
Boiling Point: 115°/1mm
MDL No.: MFCD00042112
Formula:
C₁₄H₆F₂₀O₂ MW: 586,17 g/mol Boiling Pt: 115 °C (1 mmHg) Melting Pt: 44…53 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00042112 CAS Number: 4998-38-3 EINECS: 225-659-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...