2,4,6,8-Tetramethyl-2,4,6,8-tetravinylcyclotetrasiloxane 97%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
2,4,6,8-Tetramethyl-2,4,6,8-tetravinyl-1,3,5,7,2,4,6,8-tetraoxatetrasilocane
L16645.18
L16645.30
L16645.18EA
89.1
EUR
L16645.18
L16645.30
2,4,6,8-Tetramethyl-2,4,6,8-tetravinylcyclotetrasiloxane 97%
2,4,6,8-Tetramethyl-2,4,6,8-tetravinylcyclotetrasiloxane
Purity: 97%
CAS No.: 2554-06-5
Molecular Formula: C12H24O4Si4
Formula Weight: 344.67
Flash Point: 112°(233°F)
Refractive Index: 1.4340
Melting Point: -43°
Boiling Point: 110°/10mm
Beilstein Reference: 1807272
MDL No.: MFCD00040293
Formula:
C₁₂H₂₄O₄Si₄ Boiling Pt: 110 °C (10 mmHg) Melting Pt: –43 °C Density: 0,997 Flash Pt: 112 °C (233 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00040293 CAS Number: 2554-06-5 EINECS: 219-863-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...