2,2,3,3,3-Pentafluoropropylamine 97%
Supplier: Thermo Scientific
|
Danger
|
L16819.03
L16819.06
L16819.03EA
121
EUR
L16819.03
L16819.06
2,2,3,3,3-Pentafluoropropylamine 97%
2,2,3,3,3-Pentafluoropropylamine
Purity: 97%
CAS No.: 422-03-7
Molecular Formula: CF3CF2CH2NH2
Formula Weight: 149.06
Storage Temperature: 2°C to 8°C
Flash Point:
Formula:
C₃H₄F₅N Boiling Pt: 49…50 °C Density: 1,400 Flash Pt: >100 °C (212 °F) |
MDL Number:
MFCD00042459 CAS Number: 422-03-7 EINECS: 207-010-6 ADR: 8,III |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...