Perfluoro(4-methylpent-2-ene) mixture E/Z ≥90% cont 5-10% perfluoro(2-methyl-2-pentene)

Supplier: Thermo Scientific

L16836.14 L16836.22
L16836.14EA 49.4 EUR
L16836.14 L16836.22
Perfluoro(4-methylpent-2-ene) mixture E/Z ≥90% cont 5-10% perfluoro(2-methyl-2-pentene)
Perfluoro(4-methylpent-2-ene)

Purity: 90+% CAS No.: 2070-70-4 Molecular Formula: C6F12 Formula Weight: 300.05 Refractive Index: 1.3000 MDL No.: MFCD00153253

Formula: C₆F₁₂
Storage Temperature: Ambient
MDL Number: MFCD00153253
CAS Number: 2070-70-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR