2,4,6-Tris(heptafluoropropyl)-1,3,5-triazine mass spec standard
Supplier: Thermo Scientific
Synonyms:
2,4,6-Tris(perfluoropropyl)-1,3,5-triazine
L16883.06EA
189
EUR
L16883.06
L16883.03
2,4,6-Tris(heptafluoropropyl)-1,3,5-triazine mass spec standard
2,4,6-Tris(heptafluoropropyl)-1,3,5-triazine
Formula:
C₁₂F₂₁N₃ MW: 585,12 g/mol Boiling Pt: 165 °C Density: 1,716 Storage Temperature: Ambient |
MDL Number:
MFCD00042438 CAS Number: 915-76-4 EINECS: 213-023-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...