11H-Eicosafluoroundecanoyl chloride ≥97%
Supplier: Thermo Fisher Scientific
L16896.03
L16896.06
L16896.03EA
0
EUR
L16896.03
L16896.06
11H-Eicosafluoroundecanoyl chloride ≥97%
11H-Eicosafluoroundecanoyl chloride
Purity: 97%
CAS No.: 2248-93-3
Molecular Formula: H(CF2)10COCl
Formula Weight: 564.55
Melting Point: 54-56°
UN No.: UN3261
MDL No.: MFCD00042119
Formula:
C₁₁HClF₂₀O Melting Pt: 54…56 °C |
MDL Number:
MFCD00042119 CAS Number: 2248-93-3 UN: 3261 ADR: , |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...