Ethyl-3-amino-4,4,4-trifluorobutyrate 95%
Supplier: Thermo Scientific
|
Danger
|
L17025.03
L17025.MD
L17025.03EA
160
EUR
L17025.03
L17025.MD
Ethyl-3-amino-4,4,4-trifluorobutyrate 95%
Ethyl-3-amino-4,4,4-trifluorobutyrate
Purity: 95%
CAS No.: 170804-18-9
Molecular Formula: C6H10F3NO2
Formula Weight: 185.15
Refractive Index: 1.3831
Boiling Point: 70-71°/4mm
Beilstein Reference: 6136102
UN No.: UN2735
MDL No.: MFCD01075754
Formula:
C₆H₁₀F₃NO₂ MW: 185,15 g/mol Boiling Pt: 70…71 °C (4 mmHg) Storage Temperature: Ambient |
MDL Number:
MFCD01075754 CAS Number: 170804-18-9 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...