2,4,6-Trimethylbenzeneboronic acid neopentyl glycol ester 99%
Supplier: Thermo Scientific
Synonyms:
2-Mesityl-5,5-dimethyl-1,3,2-dioxaborinane
L17232.03
L17232.06
L17232.03EA
40.7
EUR
L17232.03
L17232.06
2,4,6-Trimethylbenzeneboronic acid neopentyl glycol ester 99%
2,4,6-Trimethylbenzeneboronic acid neopentyl glycol ester
Purity: 99%
CAS No.: 214360-78-8
Molecular Formula: C14H21BO2
Formula Weight: 232.13
Melting Point: 46-48°
Beilstein Reference: 8689636
MDL No.: MFCD01318178
Formula:
C₁₄H₂₁BO₂ MW: 232,13 g/mol Melting Pt: 46…48 °C Storage Temperature: Ambient |
MDL Number:
MFCD01318178 CAS Number: 214360-78-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...