6-Methylindole-3-carboxyaldehyde 97%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
6-Methyl-1H-indole-3-carbaldehyde
L17508.03
L17508.MD
L17508.03EA
121
EUR
L17508.03
L17508.MD
6-Methylindole-3-carboxyaldehyde 97%
6-Methylindole-3-carboxyaldehyde
Purity: 98%
CAS No.: 4771-49-7
Molecular Formula: C10H9NO
Formula Weight: 159.19
Melting Point: 185-189°
Beilstein Reference: 386914
MDL No.: MFCD00049347
Formula:
C₁₀H₉NO Melting Pt: 185…189 °C Storage Temperature: Ambient |
MDL Number:
MFCD00049347 CAS Number: 4771-49-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...